Learn More
Cayman Chemical Tuftsin (human) (trifluoroacetate salt), Purity: ≥95%, 100 mg
SDP
Catalog No. 502946414
Encompass_Preferred
A phagocytosis-stimulating peptide; binds human PMN, human monocytes, and mouse macrophages (Kds = 130, 125, and 53 nM, respectively); increases phagocytosis of IgG-coated ovine red blood cells by 61% in isolated mouse peritoneal macrophages at 200 nM; increases phagocytosis of S. aureus, L. monocytogenes, and S. oviformis by isolated human, mouse, rabbit, dog, or guinea pig phagocytes from 0.01-0.15 µg/ml; increases survival in a syngeneic mouse model of 2-methylcholanthrene-induced fibrosarcoma at 0.1, 0.5, and 25 mg/kg
Catalog No. 50-294-6414 Supplier Cayman Chemical Supplier No. 33804100 MG
A phagocytosis-stimulating peptide; binds human PMN, human monocytes, and mouse macrophages (Kds = 130, 125, and 53 nM, respectively); increases phagocytosis of IgG-coated ovine red blood cells by 61% in isolated mouse peritoneal macrophages at 200 nM; increases phagocytosis of S. aureus, L. monocytogenes, and S. oviformis by isolated human, mouse, rabbit, dog, or guinea pig phagocytes from 0.01-0.15 µg/ml; increases survival in a syngeneic mouse model of 2-methylcholanthrene-induced fibrosarcoma at 0.1, 0.5, and 25 mg/kg