Learn More
Medchemexpress LLC HY-112813 5mg Medchemexpress, TUG-1375 CAS:2247372-59-2 Purity:>98%
SDP
Catalog No. 501932439
Encompass_Preferred
Medchemexpress, HY-112813 5mg TUG-1375 CAS:2247372-59-2 TUG-1375 is an agonist of free fatty acid receptor 2 ( FFA2/GPR43 ), with a pK i of 6.69. TUG-1375 is inactive on FFA3, FFA4, PPARα, PPARγ, PPARδ, LXRα or LXRβ [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2439 Supplier Medchemexpress LLC Supplier No. HY1128135MG
Medchemexpress, HY-112813 5mg TUG-1375 CAS:2247372-59-2 TUG-1375 is an agonist of free fatty acid receptor 2 ( FFA2/GPR43 ), with a pK i of 6.69. TUG-1375 is inactive on FFA3, FFA4, PPARα, PPARγ, PPARδ, LXRα or LXRβ [1] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.