Learn More
Medchemexpress LLC TUG-1387 | 6319-64-8 | 98.7% | 315.37 | C21H17NO2 | 5 MG
SDP
Catalog No. 5000262311
Encompass_Preferred
TUG-1387 is a small-molecule negative control used in studies of free fatty acid receptor 4 (FFA4). As an analog that lacks inhibitory activity, it is intended for use as an inactive comparator in functional assays and validation experiments. The compound is supplied as a high-purity solid, molecular formula C21H17NO2, molecular weight 315.37.
Catalog No. 50-002-62311 Supplier Medchemexpress LLC Supplier No. HY1291365MG
TUG-1387 is a small-molecule negative control used in studies of free fatty acid receptor 4 (FFA4). As an analog that lacks inhibitory activity, it is intended for use as an inactive comparator in functional assays and validation experiments. The compound is supplied as a high-purity solid, molecular formula C21H17NO2, molecular weight 315.37.