Learn More
Medchemexpress LLC HY-14363 1mg Medchemexpress, TUG-424 CAS:1082058-99-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY143631MG
Encompass_Preferred
Medchemexpress, HY-14363 1mg TUG-424 CAS:1082058-99-8 TUG-424 is a potent and selective free fatty acid receptor 1 (FFA1/GPR40) agonist with an EC50 of 32 nM. TUG-424 significantly increases glucose-stimulated insulin secretion at 100 nM. TUG-424 may serve to explore the role of FFA1 in metabolic diseases such as diabetes or obesity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9902