Learn More
Apexbio Technology LLC TUG-770 1402601-82-4 5mg
SDP
Catalog No. 5000095444
Encompass_Preferred
TUG-770 (CAS 1402601-82-4) is a potent agonist of the free fatty acid receptor 1 (FFA1/GPR40) a G protein-coupled receptor implicated in the enhancement of glucose-stimulated insulin secretion by pancreatic -cells TUG-770 demonstrates high affinity for FFA1 with an EC50 of 6 nM and over 150-fold selectivity compared to FFA4 as well as high selectivity over other receptors and transporters In INS-1E rat -cell lines TUG-770 promotes insulin secretion in the presence of elevated glucose Pharmacokinetic studies in mice indicate favorable oral absorption and bioavailability with dose-dependent reductions in blood glucose during glucose tolerance tests TUG-770 is a valuable tool for investigating FFA1-mediated pathways and glucose homeostasis in metabolic research
Catalog No. 50-000-95444 Supplier Apexbio Technology LLC Supplier No. A38955
TUG-770 (CAS 1402601-82-4) is a potent agonist of the free fatty acid receptor 1 (FFA1/GPR40) a G protein-coupled receptor implicated in the enhancement of glucose-stimulated insulin secretion by pancreatic -cells TUG-770 demonstrates high affinity for FFA1 with an EC50 of 6 nM and over 150-fold selectivity compared to FFA4 as well as high selectivity over other receptors and transporters In INS-1E rat -cell lines TUG-770 promotes insulin secretion in the presence of elevated glucose Pharmacokinetic studies in mice indicate favorable oral absorption and bioavailability with dose-dependent reductions in blood glucose during glucose tolerance tests TUG-770 is a valuable tool for investigating FFA1-mediated pathways and glucose homeostasis in metabolic research