Learn More
Medchemexpress LLC HY-15697 5mg Medchemexpress, TUG-770 CAS:1402601-82-4 Purity:>98%
SDP
Catalog No. 502081770
Encompass_Preferred
Medchemexpress, HY-15697 5mg TUG-770 CAS:1402601-82-4 TUG-770 is a potent, selective and orally active GPR40/FFA1 agonist with an EC50 of 6 nM for human FFA1. TUG-770 shows a high selectivity for FFA1 over FFA2, FFA3, FFA4, PPARγ, other receptors, transporters, and enzymes. TUG-770 can be uesd for type 2 diabetes research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1770 Supplier Medchemexpress LLC Supplier No. HY156975MG
Medchemexpress, HY-15697 5mg TUG-770 CAS:1402601-82-4 TUG-770 is a potent, selective and orally active GPR40/FFA1 agonist with an EC50 of 6 nM for human FFA1. TUG-770 shows a high selectivity for FFA1 over FFA2, FFA3, FFA4, PPARγ, other receptors, transporters, and enzymes. TUG-770 can be uesd for type 2 diabetes research. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.