Learn More
Selleck Chemical LLC TUG-891-5mg
SDP
Catalog No. 502389957
Encompass_Preferred
TUG-891 is a potent and selective agonist of the long chain free fatty acid (LCFA) receptor 4 (FFA4/GPR120), which demonstrates both potential opportunity and possible challenges to therapeutic agonism.
Catalog No. 50-238-9957 Supplier Selleck Chemical LLC Supplier No. S67755MG
TUG-891 is a potent and selective agonist of the long chain free fatty acid (LCFA) receptor 4 (FFA4/GPR120), which demonstrates both potential opportunity and possible challenges to therapeutic agonism.