Learn More
Apexbio Technology LLC Tulathromycin A 217500-96-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000094209
Encompass_Preferred
Tulathromycin A (CAS 217500-96-4) is a small-molecule inhibitor targeting bacterial ribosomes It is designed to inhibit protein synthesis thereby impeding bacterial growth Tulathromycin A exerts its biological activity primarily through inhibition of bacterial protein synthesis by binding to the 50S subunit of bacterial ribosomes In experimental studies Tulathromycin A demonstrates inhibitory activity against various respiratory pathogens with MIC90 values of 2 g/ml for Mannheimia haemolytica 1-2 g/ml for Pasteurella multocida 0 5-4 g/ml for Histophilus somni and 4-16 g/ml for Actinobacillus pleuropneumoniae Based on these pharmacological properties Tulathromycin A holds research potential in the study of bovine and swine respiratory disease models
Catalog No. 50-000-94209 Supplier Apexbio Technology LLC Supplier No. A38965.1
Tulathromycin A (CAS 217500-96-4) is a small-molecule inhibitor targeting bacterial ribosomes It is designed to inhibit protein synthesis thereby impeding bacterial growth Tulathromycin A exerts its biological activity primarily through inhibition of bacterial protein synthesis by binding to the 50S subunit of bacterial ribosomes In experimental studies Tulathromycin A demonstrates inhibitory activity against various respiratory pathogens with MIC90 values of 2 g/ml for Mannheimia haemolytica 1-2 g/ml for Pasteurella multocida 0 5-4 g/ml for Histophilus somni and 4-16 g/ml for Actinobacillus pleuropneumoniae Based on these pharmacological properties Tulathromycin A holds research potential in the study of bovine and swine respiratory disease models