Learn More
Medchemexpress LLC Tulisokibart (PRA023), 10 mg, Humanized IgG1-κ Monoclonal Antibody
SDP
Catalog No. NC2937923
Encompass_Preferred
Tulisokibart (PRA023) is a humanized IgG1-κ monoclonal antibody targeting to TNFSF15/TL1A. Tulisokibart can be used to study a variety of inflammatory/fibrotic diseases, such as Crohn's Disease (CD).
Catalog No. NC2937923 Supplier Medchemexpress LLC Supplier No. HYP99000710MG
Tulisokibart (PRA023) is a humanized IgG1-κ monoclonal antibody targeting to TNFSF15/TL1A. Tulisokibart can be used to study a variety of inflammatory/fibrotic diseases, such as Crohn's Disease (CD).