Learn More
Medchemexpress LLC Tulobuterol | 41570-61-0 | MFCD00867022 | 99.7% | 100 MG
SDP
Catalog No. 5000412395
Encompass_Preferred
Tulobuterol is a long-acting β2-adrenoceptor agonist that helps reduce the frequency of exacerbations of chronic obstructive pulmonary disease and bronchial asthma. This sympathomimetic agent is also used as a transdermal patch, increasing normal diaphragm muscle strength. It is intended for research use only.
  • Acts as a β2-adrenoceptor agonist
  • Reduces RV14 RNA levels and sICAM-1 concentrations in infected cells
  • Decreases acidic endosome numbers and fluorescence
  • Inhibits RV14 infection-induced secretion of inflammatory cytokines (IL-1β, IL-6, and IL-8)
  • Restores and maintains diaphragm muscle contractility
Catalog No. 50-004-12395 Supplier Medchemexpress LLC Supplier No. HYB1810100MG
Tulobuterol is a long-acting β2-adrenoceptor agonist that helps reduce the frequency of exacerbations of chronic obstructive pulmonary disease and bronchial asthma. This sympathomimetic agent is also used as a transdermal patch, increasing normal diaphragm muscle strength. It is intended for research use only.
  • Acts as a β2-adrenoceptor agonist
  • Reduces RV14 RNA levels and sICAM-1 concentrations in infected cells
  • Decreases acidic endosome numbers and fluorescence
  • Inhibits RV14 infection-induced secretion of inflammatory cytokines (IL-1β, IL-6, and IL-8)
  • Restores and maintains diaphragm muscle contractility