Learn More
Aviva Systems Biology Corp TULP2 Antibody - N-terminal region (ARP38295_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38295_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TULP2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TULP2. Alternate names: TUBL2, CT65ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-56599