Learn More
Aviva Systems Biology Corp TULP3 Antibody - middle region (ARP33473_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33473_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TULP3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human TULP3. Alternate names: MGC45295, TUBL3ApplicationWB. Species reactivity: Cow, Dog, Horse, Human, Mouse, Rabbit
Catalog No. 50-014-55037