Learn More
Aviva Systems Biology Corp TULP4 Antibody - N-terminal region (ARP39410_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39410_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against TULP4. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human TULP4. Alternate names: KIAA1397, TUSP, RP3-442A17.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57021