Learn More
Medchemexpress LLC HY-109046 10mg Medchemexpress, Tulrampator CAS:1038984-31-4 Purity:>98%
SDP
Catalog No. 501968659
Encompass_Preferred
Medchemexpress, HY-109046 10mg Tulrampator CAS:1038984-31-4 Tulrampator (CX-1632) is an orally bioavailable positive AMPAR (allosteric modulator of AMPA receptor). Antidepressant. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8659 Supplier Medchemexpress LLC Supplier No. HY10904610MG
Medchemexpress, HY-109046 10mg Tulrampator CAS:1038984-31-4 Tulrampator (CX-1632) is an orally bioavailable positive AMPAR (allosteric modulator of AMPA receptor). Antidepressant. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.