Learn More
V&P Scientific Inc STIR ELEMENT, BAR, Stainless Steel, Parylene C Coated, Used in Microplates, Tubes/Vials, 1000/pack
Catalog No. NC9273768
Encompass
STIR ELEMENT, BAR, Stainless Steel, Parylene C Coated, 3.0mm Length, 2.0mm Width, 1.7mm Height, 200°C Max Operating Temp for Coating
Catalog No. NC9273768 Supplier V&P Scientific Inc Supplier No. VP 7121
STIR ELEMENT, BAR, Stainless Steel, Parylene C Coated, 3.0mm Length, 2.0mm Width, 1.7mm Height, 200°C Max Operating Temp for Coating