Learn More
V&P Scientific Inc STIR ELEMENT, SUPER DISCS, Stainless Steel, Parylene C Coated, Used in Microplates, Syringes, Tubes/Vials, 1000/pack
Catalog No. NC0187947
Encompass
STIR ELEMENT, SUPER DISCS, Stainless Steel, Parylene C Coated, 5.1mm Diameter, 0.7mm Height, 200°C Max Operating Temp for Coating
Catalog No. NC0187947 Supplier V&P Scientific Inc Supplier No. VP 721F1
STIR ELEMENT, SUPER DISCS, Stainless Steel, Parylene C Coated, 5.1mm Diameter, 0.7mm Height, 200°C Max Operating Temp for Coating