Learn More
V&P Scientific Inc STIR ELEMENT, CYLINDER, Stainless Steel, Passivated, Used in Microplates, Tubes/Vials, 200/pack
Catalog No. NC9012173
Encompass
STIR ELEMENT, CYLINDER, Stainless Steel, Passivated, 1.9mm Diameter, 4.8mm Length, 600°C Max Operating Temp
Catalog No. NC9012173 Supplier V&P Scientific Inc Supplier No. VP 711D
STIR ELEMENT, CYLINDER, Stainless Steel, Passivated, 1.9mm Diameter, 4.8mm Length, 600°C Max Operating Temp