Learn More
V&P Scientific Inc STIR ELEMENT, CYLINDER, Stainless Steel, Parylene C Coated, Used in Microplates, Tubes/Vials, 1000/pack
Catalog No. NC9915898
Encompass
STIR ELEMENT, CYLINDER, Stainless Steel, Parylene C Coated, 2.0mm Diameter, 4.8mm Length, 200°C Max Operating Temp for Coating, Used for Parallel Synthesis/Reaction Optimization, (1000/Pack)
Catalog No. NC9915898 Supplier V&P Scientific Inc Supplier No. VP 711D1
STIR ELEMENT, CYLINDER, Stainless Steel, Parylene C Coated, 2.0mm Diameter, 4.8mm Length, 200°C Max Operating Temp for Coating, Used for Parallel Synthesis/Reaction Optimization, (1000/Pack)