Learn More
V&P Scientific Inc STIR ELEMENT, CYLINDER, Stainless Steel, Parylene C Coated, Used in Microplates, Tubes/Vials, (1,000/Pack), 1000/pack
Catalog No. NC9988699
Encompass
STIR ELEMENT, CYLINDER, Stainless Steel, Parylene C Coated, 1.3mm Diameter, 1.6mm Length, 200°C Max Operating Temp for Coating, (1000/Pack)
Catalog No. NC9988699 Supplier V&P Scientific Inc Supplier No. VP 7161
STIR ELEMENT, CYLINDER, Stainless Steel, Parylene C Coated, 1.3mm Diameter, 1.6mm Length, 200°C Max Operating Temp for Coating, (1000/Pack)