Learn More
Epigentek Inc TuMinute PCR Clean-Up Kit (100 samples)
SDP
Catalog No. 501096074
Encompass_Preferred
The TuMinute PCR Clean-Up Kit is a complete set of essential components which enables the experimenter to efficiently cleanup PCR products from various sources including conventional or real time thermal cycling PCR and isothermal gene amplification (PCR). The entire procedure can be completed within 2 minutes and is suitable for direct use in the following applications: ligation/transformation, sequencing, restriction digestion, microarray analysis, labeling, microinjection, and in vitro transcription.
Catalog No. 50-109-6074 Supplier Epigentek Inc Supplier No. P10052
The TuMinute PCR Clean-Up Kit is a complete set of essential components which enables the experimenter to efficiently cleanup PCR products from various sources including conventional or real time thermal cycling PCR and isothermal gene amplification (PCR). The entire procedure can be completed within 2 minutes and is suitable for direct use in the following applications: ligation/transformation, sequencing, restriction digestion, microarray analysis, labeling, microinjection, and in vitro transcription.