Learn More
Cusabio Technology LLC Tumor necrosis factor A TNF-A
Catalog No. 5000050122
Encompass_Preferred
This kit detects TNF-A in rats with high specificity and sensitivity designed for quantitative measurement in serum plasma cell culture supernates and tissue homogenates Utilizing a sandwich ELISA technique it offers a detection range of 6 25 pg/mL to 400 pg/mL and a sensitivity as low as 1 56 pg/mL ensuring precise analysis of low-abundance samples The assay requires 50-100 L of sample per test and delivers results within 1-5 hours depending on protocol optimization Absorbance is measured at 450 nm providing a reliable readout for TNF-A concentration Ideal for research applications requiring accurate cytokine profiling this kit combines efficiency with robust performance across diverse biological matrices
Catalog No. 50-000-50122 Supplier Cusabio Technology LLC Supplier No. CSBE11987R
This kit detects TNF-A in rats with high specificity and sensitivity designed for quantitative measurement in serum plasma cell culture supernates and tissue homogenates Utilizing a sandwich ELISA technique it offers a detection range of 6 25 pg/mL to 400 pg/mL and a sensitivity as low as 1 56 pg/mL ensuring precise analysis of low-abundance samples The assay requires 50-100 L of sample per test and delivers results within 1-5 hours depending on protocol optimization Absorbance is measured at 450 nm providing a reliable readout for TNF-A concentration Ideal for research applications requiring accurate cytokine profiling this kit combines efficiency with robust performance across diverse biological matrices