Learn More
Cusabio Technology LLC Tumor necrosis factor A TNF-A
Catalog No. 5000048949
Encompass_Preferred
This kit detects TNF-A in rat samples including tissue homogenates cell culture supernates and bronchoalveolar lavage fluid Designed for quantitative analysis using a sandwich ELISA method it provides a detection range of 15 6 pg/mL to 1000 pg/mL with a sensitivity of 3 9 pg/mL The assay requires 50-100 L of sample per test and delivers results within 1-5 hours Absorbance is measured at 450 nm ensuring reliable and precise detection of TNF-A levels for applications in inflammation research immune response studies and related biomedical fields
Catalog No. 50-000-48949 Supplier Cusabio Technology LLC Supplier No. CSBE11987RIS
Add to Cart
Add to Cart
This kit detects TNF-A in rat samples including tissue homogenates cell culture supernates and bronchoalveolar lavage fluid Designed for quantitative analysis using a sandwich ELISA method it provides a detection range of 15 6 pg/mL to 1000 pg/mL with a sensitivity of 3 9 pg/mL The assay requires 50-100 L of sample per test and delivers results within 1-5 hours Absorbance is measured at 450 nm ensuring reliable and precise detection of TNF-A levels for applications in inflammation research immune response studies and related biomedical fields