Learn More
STEMCELL Technologies Mouse Recombinant TNF-alpha, Size: 1000 µg
SDP
Supplier:  STEMCELL Technologies 78069.3
Encompass
Tumor necrosis factor-α (TNF-α) is a pro-inflammatory cytokine that activates NF-κB, MAPK, and PI3K/AKT pathways. Activated T cells and macrophages are the primary producers of TNF-α in response to inflammation and infectious conditions. Many other cell types have been shown to produce TNF-α, among them B cells, NK cells, mast cells, neutrophils, dendritic cells, microglia, endothelial cells, smooth muscle cells, cardiomyocytes, and fibroblasts. TNF-α has cytotoxic effects on cancer cells in vitro by stimulating anti-tumor immunosuppressive responses. TNF-α stimulates expression of E- and P-selectins, thus facilitating adhesion of neutrophils, monocytes, and memory T cells to activated platelets and endothelial cells (Zelová & Hosek). Other effects of TNF-α include vasodilatation and edema formation.
Catalog No. 50-226-9646