Learn More
Biomatik Corporation Tumor Necrosis Factor Alpha (Tnfa) Polyclonal Antibody
Supplier:  Biomatik Corporation CAU30311
Encompass_Preferred
Tumor Necrosis Factor Alpha (Tnfa) Polyclonal Antibody is a Mouse (Mus musculus) Polyclonal Antibody to Tumor Necrosis Factor Alpha (TNFa). The Tumor Necrosis Factor Alpha (Tnfa) Polyclonal Antibody has been validated for the following applications: WB, IHC, ICC, IP. Host: Rabbit. Species: Mouse (Mus musculus). Immunogen: Leu80~Leu235. Applications: WB, IHC, ICC, IP. Target Synonyms: Tumor Necrosis Factor Alpha (TNFa). Accession Number: P06804. Gene ID: 21926. Storage: 2°C to 8°C for frequent use, -20°C for 12 months. Avoid repeated freeze/thaw cycles. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4786