Learn More
Cusabio Technology LLC Tumor necrosis factor B TNF-B
Catalog No. 5000050743
Encompass_Preferred
This kit detects TNF-B in rat serum plasma and tissue homogenates with a detection range of 7 8 pg/mL to 500 pg/mL offering a sensitivity of 1 95 pg/mL Utilizing a quantitative sandwich ELISA method it requires a sample volume of 50-100 L and delivers results within 1-5 hours The assay measures absorbance at 450 nm ensuring reliable and precise quantification of TNF-B levels in various biological samples for research applications
Catalog No. 50-000-50743 Supplier Cusabio Technology LLC Supplier No. CSBE07379R
This kit detects TNF-B in rat serum plasma and tissue homogenates with a detection range of 7 8 pg/mL to 500 pg/mL offering a sensitivity of 1 95 pg/mL Utilizing a quantitative sandwich ELISA method it requires a sample volume of 50-100 L and delivers results within 1-5 hours The assay measures absorbance at 450 nm ensuring reliable and precise quantification of TNF-B levels in various biological samples for research applications