Learn More
Sigma Aldrich Fine Chemicals Biosciences TUNEL Enzyme from calf thy
Catalog No. 502891565
Encompass
TUNEL enzyme solution, containing terminal deoxynucleotidyl transferase from calf thymus recombinant in E. coli.One of the two components required for performing the TUNEL reaction to detect apoptosis in situ.
Catalog No. 50-289-1565 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11767305001
TUNEL enzyme solution, containing terminal deoxynucleotidyl transferase from calf thymus recombinant in E. coli.One of the two components required for performing the TUNEL reaction to detect apoptosis in situ.