Learn More
Sigma Aldrich Fine Chemicals Biosciences TUNEL Label Mix
Catalog No. 502900237
Encompass
The nucleotide-labeling mix (TUNEL Label) contains fluorescein-dUTP and -dNTPs both needed to perform the TUNEL (terminal deoxynucleotidyl transferase dUTP nick end labeling) reaction for the detection of apoptosis in situ. The nucleotide-labeling mix is used in combination with the TUNEL enzyme to prepare a TUNEL reaction mixture. This reaction mixture is used to label DNA-strand breaks for detecting and quantifying apoptotic cell death at a single-cell level in cells and tissues.
Catalog No. 50-290-0237 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 11767291910
The nucleotide-labeling mix (TUNEL Label) contains fluorescein-dUTP and -dNTPs both needed to perform the TUNEL (terminal deoxynucleotidyl transferase dUTP nick end labeling) reaction for the detection of apoptosis in situ. The nucleotide-labeling mix is used in combination with the TUNEL enzyme to prepare a TUNEL reaction mixture. This reaction mixture is used to label DNA-strand breaks for detecting and quantifying apoptotic cell death at a single-cell level in cells and tissues.