Learn More
Electron Microscopy Sciences EMS Tungsten Boat
SDP
Catalog No. 5033188
Encompass_Preferred
Thickness of tungsten foil is 0.05mm (0.002"), overall length is 32mm (1 1/4"), the trough: 12.7mm(L) x 9.5mm(W) x 3.2mm Deep (1/2" x 5⁄8" x 1⁄32")
Catalog No. 50-331-88 Supplier Electron Microscopy Sciences Supplier No. 73816
Thickness of tungsten foil is 0.05mm (0.002"), overall length is 32mm (1 1/4"), the trough: 12.7mm(L) x 9.5mm(W) x 3.2mm Deep (1/2" x 5⁄8" x 1⁄32")