Learn More
Electron Microscopy Sciences EMS Tungsten Boat B-2
SDP
Supplier:  Electron Microscopy Sciences 73816B2
Encompass
Thickness of tungsten foil is 0.25 mm (0.010"). Overall length is 101.6mm x 19.05mm (4" x 3⁄4"), trough: 39.37mm (L) x 2.79mm Deep, 0.92 ml
Catalog No. 50-331-89