Learn More
Electron Microscopy Sciences EMS Tungsten Boat B-3
SDP
Supplier:  Electron Microscopy Sciences 73816B3
Encompass
Thickness of tungsten foil is 0.25 mm (0.010"). Overall length is 101.6mm x 19.05mm (4" x 3⁄4"), trough: 39.1mm(L) x 6.35mm(W) x 3.3mm Deep, 0.5ml
Catalog No. 50-331-90