Learn More
Electron Microscopy Sciences EMS Tungsten Boat B-4
SDP
Catalog No. 5033194
Encompass
Thickness of tungsten foil is 0.25 mm (0.010"). Overall length is 100.5mm x 19.05mm (3.5" x 3⁄4"), trough: 28.19mm(L) x 15.87(W) x 3.3mm Deep, 0.99ml
Catalog No. 50-331-94 Supplier Electron Microscopy Sciences Supplier No. 73816B4
Thickness of tungsten foil is 0.25 mm (0.010"). Overall length is 100.5mm x 19.05mm (3.5" x 3⁄4"), trough: 28.19mm(L) x 15.87(W) x 3.3mm Deep, 0.99ml