Learn More
Electron Microscopy Sciences EMS Tungsten Boat B-5
SDP
Supplier:  Electron Microscopy Sciences 73816B5
Encompass
Thickness of tungsten foil is 0.25 mm (0.010"). Overall length is 101.6mm x 12.7mm (4" x 1/2"), trough: 9.65mm Diameter x 3.81mm Deep, 0.18ml
Catalog No. 50-331-93