Learn More
Electron Microscopy Sciences EMS Tungsten Boat B-6
SDP
Supplier:  Electron Microscopy Sciences 73816B6
Encompass
Thickness of tungsten foil is 0.25 mm (0.010"). Overall length is 101.6mm x 39.1mm(L) (4" x 1/2"), trough: 39.1mm(L) x 12mm(W) x 3.55mm Deep, 1.0ml
Catalog No. 50-331-92