Learn More
Electron Microscopy Sciences EMS Tungsten Boat B-7
SDP
Supplier:  Electron Microscopy Sciences 73816B7
Encompass
Thickness of tungsten foil is 0.25 mm (0.010"). Overall length is 101.6mm x 25.4mmL x (4" x 1"), trough: 53.84mm(L)x25mm(W) x2.28mm Deep, 2.43ml
Catalog No. 50-331-91