Learn More
Braintree Scientific Tungsten Carbide Iris Scissors, straight, 4.5""""
SDP
Catalog No. 501955882
Encompass
Tungsten Carbide Iris Scissors. A unique tungsten carbide cutting edge provide effortless yet accurate cuts through the toughest tissue. Shanks are tempered to allow proper flexibility and comfort. Finger rings are well rounded and polished allowing better feel while preventing finger fatigue. Tempered and hardened for a longer sharper life. The SuperCut blade has a specially designed razor sharp edge. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5882 Supplier Braintree Scientific Supplier No. SCT405
Tungsten Carbide Iris Scissors. A unique tungsten carbide cutting edge provide effortless yet accurate cuts through the toughest tissue. Shanks are tempered to allow proper flexibility and comfort. Finger rings are well rounded and polished allowing better feel while preventing finger fatigue. Tempered and hardened for a longer sharper life. The SuperCut blade has a specially designed razor sharp edge. **Sold for research purposes only and not for human use**