Learn More
TOSCO Tungsten Carbide Pencil
Catalog No. 13378
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
13-378 Each
1 options
This item is not returnable. View return policy
Catalog No. 13-378 Supplier TOSCO Supplier No. SK63971
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Marks glass and metal
Stainless-steel pencil with tungsten carbide point and protective cap.
Specifications
Product Type Tungsten Carbide Pencil
Material Stainless Steel
Tip Material Tungsten Carbide
Includes Steel cap
For Use With (Application) Used for writing on glass and metal
Quantity Each