Learn More
Hampton Research Useful for scribing, cutting and removal of the upper coverglass that seals the LCP Sandwich Set.
SDP
Catalog No. NC1175813
Encompass
Useful for scribing, cutting and removal of the upper coverglass that seals the LCP Sandwich Set.
Catalog No. NC1175813 Supplier Hampton Research Supplier No. HR4702
Useful for scribing, cutting and removal of the upper coverglass that seals the LCP Sandwich Set.