Learn More
Hampton Research Tungsten Carbide Scribe Tip, 3/pk
SDP
Catalog No. NC1294384
Encompass
Useful for scribing, cutting and removal of the upper coverglass that seals the LCP Sandwich Set.
Catalog No. NC1294384 Supplier Hampton Research Supplier No. HR4703
Useful for scribing, cutting and removal of the upper coverglass that seals the LCP Sandwich Set.