Learn More
Fine Science Tools Strabismus Scissors, Tungsten Carbide, 11 cm
SDP
Catalog No. NC0835335
Encompass
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 22mm/ Alloy / Material: Tungsten Carbide/ Length: 11cm/ Serrated: No
Catalog No. NC0835335 Supplier Fine Science Tools Supplier No. 1457411
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 22mm/ Alloy / Material: Tungsten Carbide/ Length: 11cm/ Serrated: No