Learn More
Leica Biosystems Leica TC-65 Tungsten Carbide Disposable Blade - fits Leica cryostats (5 blades/pack)
Catalog No. NC0063355
Encompass_Preferred
Leica TC-65 tungsten carbide disposable blades provide durable, precision cutting for cryostat sectioning of hard or difficult-to-cut specimens. Constructed from tungsten carbide for extended edge life, these disposable blades are intended for cryostat use and are supplied in small multi-blade packs for routine histology workflows.
  • Tungsten carbide construction for long-lasting sharpness.
  • Dimensions 49 mm x 8 mm.
  • Designed for cryostat sectioning and routine histology.
  • Disposable design reduces cross-contamination risk.
  • Compatible with standard cryostat blade holders.
Catalog No. NC0063355 Supplier Leica Biosystems Supplier No. 14021626379
Leica TC-65 tungsten carbide disposable blades provide durable, precision cutting for cryostat sectioning of hard or difficult-to-cut specimens. Constructed from tungsten carbide for extended edge life, these disposable blades are intended for cryostat use and are supplied in small multi-blade packs for routine histology workflows.
  • Tungsten carbide construction for long-lasting sharpness.
  • Dimensions 49 mm x 8 mm.
  • Designed for cryostat sectioning and routine histology.
  • Disposable design reduces cross-contamination risk.
  • Compatible with standard cryostat blade holders.