Learn More
Strem, An Ascensus Company CAS# 14040-11-0. 100g. Tungsten carbonyl, 99% (<0.3%-Mo). MFCD00011462
SDP
Catalog No. 5090113021
Encompass
CAS# 14040-11-0. 100g. Tungsten carbonyl, 99% (<0.3%-Mo). MFCD00011462. Molecular Weight: 351.92. Molecular Formula: W(CO)6. Color/form: white xtl. Strem# 74-2200. http://www.strem.com/catalog/v/74-2200/
Catalog No. 50-901-13021 Supplier Strem, An Ascensus Company Supplier No. 742200100G
CAS# 14040-11-0. 100g. Tungsten carbonyl, 99% (<0.3%-Mo). MFCD00011462. Molecular Weight: 351.92. Molecular Formula: W(CO)6. Color/form: white xtl. Strem# 74-2200. http://www.strem.com/catalog/v/74-2200/