Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tungsten disulfide, 99%, particle size 0.7-1.1 micrometer, Thermo Scientific™

Catalog No. AC222765000
Change view
Click to view available options
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC222765000 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC222765000 Supplier Thermo Scientific Chemicals Supplier No. 222765000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 12138-09-9
Quantity 500g
Assay Percent Range 99%
Molecular Formula S2W
Linear Formula WS2
MDL Number MFCD00011467
Synonym tungsten disulfide, tungsten sulfide ws2, tungsten disulphide, tungsten iv sulfide, bis sulfanylidene tungsten, dithioxotungsten, tungstenite, disulfidotungsten, monotungsten disulfide, ws2
InChI Key ITRNXVSDJBHYNJ-UHFFFAOYSA-N
SMILES S=[W]=S
IUPAC Name bis(sulfanylidene)tungsten
Molecular Weight (g/mol) 247.96
PubChem CID 82938
ChEBI CHEBI:30521
Formula Weight 247.98
Percent Purity ≥98.5%
Packaging Glass bottle
Density 7.5000g/mL
Chemical Name or Material Tungsten disulfide, 99%
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 235-243-3
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.