Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tungsten Disulfide Quantum Dots

Catalog No. 91429020ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
91-429-020ML 20 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 91-429-020ML Supplier Sigma Aldrich Supplier No. 91429020ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The transformation of 2D materials in QDs form supplement excellent properties from their quantum confinement effects, surface-volume ratio, solubility, edge-effects, tunable PL, ease to functionalize and fluorescence properties. These 2D QDs are demanding to impart new dimension to the 2D nanosheets and in order to advance to next level, the better understanding of the properties, synthesis methods and applications of 2D QDs are needed. The exciting optical properties of the WS2 QDs such as strong excitonic absorption bands covering the visible region, excellent excitonic fluorescence, highly tunable PL, atomically-thin structure, molecular size, biocompatibility, easy functionalization, enhanced photostability, higher molar extinction coefficients and long-term photostability make them attractive candidates for optical sensing.

Specifications

Quantity 20 mL
Molecular Formula WS2
Linear Formula WS2
MDL Number MFCD00011467
Molecular Weight (g/mol) 247.97
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.