Learn More
Analytical Sales & Services Tungsten Electrodes
Catalog No. 5000806276
Encompass
Tungsten Electrodes\n\nDiameter: 0.064”\nLength: 1.23”\n\nQty/Pkg: 25
Catalog No. 50-008-06276 Supplier Analytical Sales & Services Supplier No. 700875
Tungsten Electrodes\n\nDiameter: 0.064”\nLength: 1.23”\n\nQty/Pkg: 25