Learn More
Perkin Elmer US LLC Vis Lamp Complete, Tungsten
Catalog No. 509042196
Encompass_Preferred
For legacy LAMBDA spectrometers, PerkinElmer supplies reliable, long lasting deuterium and tungsten-halogen light sources. We recommend these replacement bulbs be installed by a PerkinElmer Service Engineer since optimal spectrometer performance depends upon the lamp’s precise alignment.
Catalog No. 50-904-2196 Supplier Perkin Elmer US LLC Supplier No. B0114620
For legacy LAMBDA spectrometers, PerkinElmer supplies reliable, long lasting deuterium and tungsten-halogen light sources. We recommend these replacement bulbs be installed by a PerkinElmer Service Engineer since optimal spectrometer performance depends upon the lamp’s precise alignment.