Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tungsten(IV) sulfide

Catalog No. 7905835G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
79-058-35G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 79-058-35G Supplier Sigma Aldrich Supplier No. 7905835G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tungsten sulfide possesses a pseudo 2 dimensional structure with applications in the Petrochemical industry as a desulfurization catalyst. Tungsten Sulfide nanoparticles have drawn research interest due to their potential in solid-state lubricants; coatings and photocatalysts.

Specifications

CAS 12138-09-9
Quantity 5 g
Molecular Formula S2W
Linear Formula WS2
MDL Number MFCD00011467
Synonym Monotungsten disulfide; Tungsten disulfide
Molecular Weight (g/mol) 247.97
Percent Purity 99% trace metals basis
Density 7.5 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.