Learn More
Analytical Sales & Services Tungsten Lamp for Beckman Mode
Catalog No. 5000807259
Encompass
Tungsten Lamp for Beckman Models 5240 DU-6 7 8 20 30 40 50; Shimadzu Models BIOSPEC 1601 Series 160 UV-12055 65 75 160U 260 265 1501 1601 1601PC 1700 UV-2501PC 3101PC
Qty/Pkg: EACH
Catalog No. 50-008-07259 Supplier Analytical Sales & Services Supplier No. 12840
Tungsten Lamp for Beckman Models 5240 DU-6 7 8 20 30 40 50; Shimadzu Models BIOSPEC 1601 Series 160 UV-12055 65 75 160U 260 265 1501 1601 1601PC 1700 UV-2501PC 3101PC
Qty/Pkg: EACH