Learn More
Analytical Sales & Services Tungsten Lamp for Beckman Mode
Catalog No. 5000804948
Encompass
Tungsten Lamp for Beckman Models DU-60 62 63 64 65 68 70 600 640 650 800 7000 7400 7500; Shimadzu Models SPD-M10AVP UV-1201 1240 1240V
Qty/Pkg: EACH
Catalog No. 50-008-04948 Supplier Analytical Sales & Services Supplier No. 12835
Tungsten Lamp for Beckman Models DU-60 62 63 64 65 68 70 600 640 650 800 7000 7400 7500; Shimadzu Models SPD-M10AVP UV-1201 1240 1240V
Qty/Pkg: EACH