Learn More
Mettler Toledo™ Tungsten Lamp Set Easy VIS
Catalog No. 01911521 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Type:
Tungsten Halogen Lamp
Catalog No. Type
01-911-521 Tungsten Halogen Lamp
1 options
This item is not returnable. View return policy
Catalog No. 01-911-521 Supplier Mettler Toledo™ Supplier No. 30705148
Add to Cart
Add to Cart
This item is not returnable. View return policy
Mettler Toledo Easy VIS Tungsten Lamps measure the absorption of visible light in a sample from 390 to 800 nm, usually contained in a cuvette. The spectrophotometer constantly monitors the lamp and alerts the user when it needs replaced, which only takes two minutes.
  • Compatible with Mettler Toledo Easy Vis Spectrophotometers
  • The Easy Vis Spectrophotometer monitors and records the lamp's usage and alerts the user well in advance when a replacement is needed
  • Easily replace your Tungsten Lamp Set in just two minutes by using the provided tool to open the two screws and place the new lamp inside
TRUSTED_SUSTAINABILITY
Specifications
Type Tungsten Halogen Lamp
Voltage 120 V