Learn More
Roboz Surgical TUNGSTEN NEEDLE 0.25 MM 10/P
SDP
Catalog No. NC0174212
Encompass
These fine needles made of hard tungsten have a 1 um tip diameter. They can be used for micro dissection at the finest level. The material is much harder than stainless steel therefore it offers increased stability with less spring action.
Catalog No. NC0174212 Supplier Roboz Surgical Supplier No. RS6064
These fine needles made of hard tungsten have a 1 um tip diameter. They can be used for micro dissection at the finest level. The material is much harder than stainless steel therefore it offers increased stability with less spring action.